Skip to Content
Products
Cart
0
US
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Tartrazine Solution
Products
Shipping
Ships Today
Product Category
histology special stains
Brand
Sigma-Aldrich
Physical Form
Quality Segment
Application
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
HT3028
Tartrazine Solution,
Expand
View Pricing
HT3024
Tartrazine Solution,
Expand
View Pricing
An unknown error has occured.