Skip to Content
MilliporeSigma
TISAB III solution

TISAB III solution

TISAB III solution
Synonym(s):
Total ionic strength adjustment buffer III solution

Products

Product Category
Brand
Physical Form
Quality Segment
Technique
Showing 1-2 of 2