Skip to Content
MilliporeSigma
TRIS borate – EDTA buffer solution for Northern and Southern blotting, solution

TRIS borate – EDTA buffer solution

TRIS borate – EDTA buffer solution for Northern and Southern blotting, solution
Synonym(s):
TBE buffer

Products

Shipping
Product Category
Brand
Physical Form
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1