
Trypsin from porcine pancreas

Synonym(s):
Porcine Trypsin, Trypsin for Mass Spectropetry
EC Number:
Products
Product Category
Brand
Biological Source
Physical Form
Product Line
Tissue
Quality Segment
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing