
Vitamin D3 solution

Synonym(s):
Vitamin D3 solution
Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64
Products
Product Category
Brand
Color
Manufacturer
Physical Form
Purity
Quality Segment
Application
Special Grade
Technique
Showing 1-2 of 2