MilliporeSigma
Mutation analysis of PTEN/MMAC1 in acute myeloid leukemia.
American journal of hematology (2000-03-08)
T C Liu, P M Lin, J G Chang, J P Lee, T P Chen, S F Lin
ABSTRACT
Recently, a putative tumor suppressor gene, PTEN/MMAC1, has been identified at chromosome 10q23.3, which encodes a 403 amino acid dual-specificity phosphatase containing a region of homology to tensin and auxillin. Somatic mutations of the PTEN/MMAC1 gene have been identified in a number of cancer cell lines and primary cancers. Mutations in PTEN/MMAC1 are most frequently found in advanced cancers. To evaluate the role of the PTEN/MMAC1 gene in leukemia, bone marrow and/or peripheral blood from 62 acute myeloid leukemia (AML) patients, 5 hemopoietic cell lines (HL60, U937, Raji, KG-1, K562), and 30 normal controls were analyzed. The results showed aberrant PTEN/MMAC1 transcripts in 15 of the 62 (24%) AML patients, 4 of the 5 cell lines (80%), and 4 of the 30 (13%) normal controls. As in our previous study of TSG101, the abnormal transcripts may result from aberrant RNA splicing as evidenced by the presence of both these aberrant transcripts and normal full length transcripts in all specimens examined. Loss of heterozygosity (LOH) analysis and PCR-SSCP of the entire coding region showed that none of the AML cases had LOH or mutation. Only one frameshift mutation at codon 130 (insertion of CCCG) with premature termination of coding sequence was observed in the U937 cell line. Our results indicate that the PTEN/MMAC1 gene may play a role in a small percentage of AML, but its significance needs to be further evaluated.