MilliporeSigma
  • The absence of the luxS gene increases swimming motility and flagella synthesis in Escherichia coli K12.
The absence of the luxS gene increases swimming motility and flagella synthesis in Escherichia coli K12.
Biochemical and biophysical research communications (2010-09-30)
Hua Ling, Aram Kang, Mui Hua Tan, Xiaobao Qi, Matthew Wook Chang
ABSTRACT
Despite the significant role of S-ribosylhomocysteinase (LuxS) in the activated methyl cycle pathway and quorum sensing, the connectivity between luxS and other cellular functions remains incomplete. Herein, we show that luxS deletion significantly increases swimming motility and flagella synthesis in Escherichia coli K12 using motility, transcriptome, and scanning electron microscopy assays. Further, based on the transcriptome and network component analyses, and known regulatory relations, we propose a conceptual genetic regulatory network underlying the increased flagella synthesis in response to luxS deletion.