MilliporeSigma
  • Adeno-associated viral vector-induced overexpression of neuropeptide Y Y2 receptors in the hippocampus suppresses seizures.
Adeno-associated viral vector-induced overexpression of neuropeptide Y Y2 receptors in the hippocampus suppresses seizures.
Brain : a journal of neurology (2010-08-07)
David P D Woldbye, Mikael Angehagen, Casper R Gøtzsche, Heidi Elbrønd-Bek, Andreas T Sørensen, Søren H Christiansen, Mikkel V Olesen, Litsa Nikitidou, Thomas V O Hansen, Irene Kanter-Schlifke, Merab Kokaia
ABSTRACT
Gene therapy using recombinant adeno-associated viral vectors overexpressing neuropeptide Y in the hippocampus exerts seizure-suppressant effects in rodent epilepsy models and is currently considered for clinical application in patients with intractable mesial temporal lobe epilepsy. Seizure suppression by neuropeptide Y in the hippocampus is predominantly mediated by Y2 receptors, which, together with neuropeptide Y, are upregulated after seizures as a compensatory mechanism. To explore whether such upregulation could prevent seizures, we overexpressed Y2 receptors in the hippocampus using recombinant adeno-associated viral vectors. In two temporal lobe epilepsy models, electrical kindling and kainate-induced seizures, vector-based transduction of Y2 receptor complementary DNA in the hippocampus of adult rats exerted seizure-suppressant effects. Simultaneous overexpression of Y2 and neuropeptide Y had a more pronounced seizure-suppressant effect. These results demonstrate that overexpression of Y2 receptors (alone or in combination with neuropeptide Y) could be an alternative strategy for epilepsy treatment.
Sigma-Aldrich
Guanosine 5′-[β-thio]diphosphate trilithium salt, ≥85% (HPLC), powder
Sigma-Aldrich
Kainic acid monohydrate, ≥98% (HPLC), from Digenea simplex