MilliporeSigma
  • Water-insoluble polymer?clay nanocomposite ion exchange resin based on N-methyl-D-glucamine ligand groups for arsenic removal.
Water-insoluble polymer?clay nanocomposite ion exchange resin based on N-methyl-D-glucamine ligand groups for arsenic removal.
Reactive functional Polymers (2012)
Urbano BF