MilliporeSigma
  • Nuclear GSK3β induces DNA double-strand break repair by phosphorylating 53BP1 in glioblastoma.
Nuclear GSK3β induces DNA double-strand break repair by phosphorylating 53BP1 in glioblastoma.
International journal of oncology (2018-01-13)
Yong Yang, Tiantian Lei, Suya Du, Rongsheng Tong, Hailian Wang, Jiao Yang, Juan Huang, Minghan Sun, Yi Wang, Zhi Dong
ABSTRACT
Glioblastoma is the most malignant and lethal subtype brain tumors with high risk of recurrence and therapeutic resistance. Emerging evidence has indicated that glycogen synthesis kinase 3 (GSK3)β plays oncogenic roles in multiple tumor types; however, the underlying mechanisms remain largely unknown. It has also been demonstrated that p53 binding protein 1 (53BP1) plays a central role in DNA double-strand break (DSB) repair. This study aimed to reveal the significance of GSK3β translocation from the cytoplasm to the nucleus, and to determine whether GSK3β induces DNA DSB repair in the nuclei of glioblastoma cells via phospho-53BP1. By performing in vitro experiments, we found that GSK3β translocated from the cytoplasm to the nucleus, and it then bound to 53BP1 following exposure to IR (IR). In addition, 53BP1-mediated DNA DSB repair was observed to be abrogated by the inhibition of GSK3β. Further experiments on the phosphorylation site of 53BP1 by GSK3β revealed that the S/T-Q motif may play a critical role. Importantly, our in vivo and in vitro data clearly indicated that GSK3β induced the phosphorylation of 53BP1 at the Ser166 site. Moreover, brain tumor xenograft models revealed that following exposure to IR plus SB216763, a specific GSK3β inhibitor, tumor growth was markedly inhibited and the survival of mice markedly increased. Based on these results, we concluded that the phosphorylation of 53BP1 by GSK3β was indispensable for DNA DSB repair. Our study also suggested that the inhibition of GSK3β by SB216763 significantly inhibited the proliferation and induced the apoptosis of glioblastoma cells. Taken together, our data indicate that GSK3β, a key phosphorylation protein for 53BP1, may be a potential target for enhancing the sensitivity of glioblastoma cells to radiation.
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection