MilliporeSigma
  • The IL-33-PIN1-IRAK-M axis is critical for type 2 immunity in IL-33-induced allergic airway inflammation.
The IL-33-PIN1-IRAK-M axis is critical for type 2 immunity in IL-33-induced allergic airway inflammation.
Nature communications (2018-04-25)
Morris Nechama, Jeahoo Kwon, Shuo Wei, Adrian Tun Kyi, Robert S Welner, Iddo Z Ben-Dov, Mohamed S Arredouani, John M Asara, Chun-Hau Chen, Cheng-Yu Tsai, Kyle F Nelson, Koichi S Kobayashi, Elliot Israel, Xiao Zhen Zhou, Linda K Nicholson, Kun Ping Lu
ABSTRACT
Interleukin 33 (IL-33) is among the earliest-released cytokines in response to allergens that orchestrate type 2 immunity. The prolyl cis-trans isomerase PIN1 is known to induce cytokines for eosinophil survival and activation by stabilizing cytokines mRNAs, but the function of PIN1 in upstream signaling pathways in asthma is unknown. Here we show that interleukin receptor associated kinase M (IRAK-M) is a PIN1 target critical for IL-33 signaling in allergic asthma. NMR analysis and docking simulations suggest that PIN1 might regulate IRAK-M conformation and function in IL-33 signaling. Upon IL-33-induced airway inflammation, PIN1 is activated for binding with and isomerization of IRAK-M, resulting in IRAK-M nuclear translocation and induction of selected proinflammatory genes in dendritic cells. Thus, the IL-33-PIN1-IRAK-M is an axis critical for dendritic cell activation, type 2 immunity and IL-33 induced airway inflammation.
Sigma-Aldrich
Anti-IRAK-M antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.