MilliporeSigma
  • Expression and subcellular localisation of AID and APOBEC3 in adenoid and palatine tonsils.
Expression and subcellular localisation of AID and APOBEC3 in adenoid and palatine tonsils.
Scientific reports (2018-01-19)
Noriko Seishima, Satoru Kondo, Kousho Wakae, Naohiro Wakisaka, Eiji Kobayashi, Makoto Kano, Makiko Moriyama-Kita, Yosuke Nakanishi, Kazuhira Endo, Tomoko Imoto, Kazuya Ishikawa, Hisashi Sugimoto, Miyako Hatano, Takayoshi Ueno, Miki Koura, Koichi Kitamura, Masamichi Muramatsu, Tomokazu Yoshizaki
ABSTRACT
Activation-induced cytidine deaminase (AID) and apolipoprotein B mRNA-editing catalytic polypeptide 3 (A3) family are cytidine deaminases that play critical roles in B-cell maturation, antiviral immunity and carcinogenesis. Adenoids and palatine tonsils are secondary lymphoid immune organs, in which AID and A3s are thought to have several physiological or pathological roles. However, the expression of AID or A3s in these organs has not been investigated. Therefore, we investigated the expression profiles of AID and A3s, using 67 samples of adenoids and palatine tonsils from patients, with reverse transcription quantitative polymerase chain reaction (RT-qPCR) and immunohistochemical analyses. AID and A3s expression levels in the adenoids and the palatine tonsils of the same individual significantly correlated with each other. Of note, AID expression level in the adenoids negatively correlated with the age (r = -0.373, P = 0.003). The younger group with adenoid vegetation and tonsillar hypertrophy showed more abundant AID expression than the older group with recurrent tonsillitis and peritonsillar abscesses (P = 0.026). Moreover, immunohistochemical analysis revealed the distribution of AID and A3s in the epithelial cells as well as germinal centres. The localisation of AID expression and its relation to age may contribute to adenoid vegetation and inflammation.
Sigma-Aldrich
Anti-APOBEC3D antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution