MilliporeSigma
  • Digestion-ligation-only Hi-C is an efficient and cost-effective method for chromosome conformation capture.
Digestion-ligation-only Hi-C is an efficient and cost-effective method for chromosome conformation capture.
Nature genetics (2018-04-28)
Da Lin, Ping Hong, Siheng Zhang, Weize Xu, Muhammad Jamal, Keji Yan, Yingying Lei, Liang Li, Yijun Ruan, Zhen F Fu, Guoliang Li, Gang Cao
ABSTRACT
Chromosome conformation capture (3C) technologies can be used to investigate 3D genomic structures. However, high background noise, high costs, and a lack of straightforward noise evaluation in current methods impede the advancement of 3D genomic research. Here we developed a simple digestion-ligation-only Hi-C (DLO Hi-C) technology to explore the 3D landscape of the genome. This method requires only two rounds of digestion and ligation, without the need for biotin labeling and pulldown. Non-ligated DNA was efficiently removed in a cost-effective step by purifying specific linker-ligated DNA fragments. Notably, random ligation could be quickly evaluated in an early quality-control step before sequencing. Moreover, an in situ version of DLO Hi-C using a four-cutter restriction enzyme has been developed. We applied DLO Hi-C to delineate the genomic architecture of THP-1 and K562 cells and uncovered chromosomal translocations. This technology may facilitate investigation of genomic organization, gene regulation, and (meta)genome assembly.
Sigma-Aldrich
Ethylene glycol-bis(succinic acid N-hydroxysuccinimide ester), powder