MilliporeSigma
  • Association of vascular indices with novel circulating biomarkers as prognostic factors for cardiovascular complications in patients with type 2 diabetes mellitus.
Association of vascular indices with novel circulating biomarkers as prognostic factors for cardiovascular complications in patients with type 2 diabetes mellitus.
Clinical biochemistry (2017-12-31)
Katerina K Naka, Katerina Papathanassiou, Aris Bechlioulis, Konstantinos Pappas, Stelios Tigas, Dimitrios Makriyiannis, Sophia Antoniou, Nikolaos Kazakos, Alexandra Margeli, Ioannis Papassotiriou, Agathocles Tsatsoulis, Lampros K Michalis
ABSTRACT
The pathophysiology of atherosclerosis in type 2 diabetes mellitus (T2DM) is multifactorial. The association of vascular indices with circulating biomarkers of inflammation and insulin resistance and their role in the long-term cardiovascular prognosis in T2DM patients were currently investigated. Patients with T2DM and poor glycemic control without known cardiovascular diseases (n=119) at baseline were enrolled and followed for about 9years. The end-point was the occurrence of any cardiovascular event (coronary heart disease, stroke, peripheral artery disease or cardiovascular death). Aortic pulse wave velocity (PWV), augmentation index (AIx), brachial flow-mediated dilation (FMD), hsCRP, Chitinase-3-like protein 1 (YKL-40), Neutrophil Gelatinase-Associated Lipocalin (NGAL), Fatty Acid Binding Protein (FABP-4) were assessed. Higher YKL-40 and NGAL were associated with higher PWV, while higher YKL-40 and FABP-4 were related to higher AIx (p<0.05 for all). In univariate Cox regression analysis, PWV>10m/s, YKL-40>78ng/ml and NGAL>42ng/ml were associated with cardiovascular events (p<0.05 for all). In multivariate analysis, after adjusting for classical risk factors and glycemic control, increased NGAL, YKL-40 and PWV and decreased FMD (i.e. ≤2.2%) (p<0.05 for all) were independently associated with cardiovascular events. In T2DM patients without established cardiovascular disease, novel indices of vascular inflammation (NGAL and YKL-40) were associated with subclinical atherosclerosis (arterial stiffness) but also with adverse clinical prognosis. Arterial stiffness and endothelial dysfunction were also independently related to adverse prognosis.
Sigma-Aldrich
Human CHI3L1 / Chitinase-3-like Protein 1 ELISA Kit