MilliporeSigma
  • Etched poly(ether ether ketone) jacket stir bar with detachable dumbbell-shaped structure for stir bar sorptive extraction.
Etched poly(ether ether ketone) jacket stir bar with detachable dumbbell-shaped structure for stir bar sorptive extraction.
Journal of chromatography. A (2018-04-24)
Wei Zhou, Chenlu Wang, Xuemei Wang, Zilin Chen
ABSTRACT
Development of stir bar sorptive extraction (SBSE) device with high stability and extraction efficiency is critical and challenging by date. In this work, etched poly(ether ether ketone) (PEEK) tube with high mechanical strength and large specific surface area was used as jacket for SBSE device. By etching with concentrated sulfuric acid, the smooth outer surface of PEEK become porous with plenty of micro holes, which was beneficial for coating of sorbents and significantly improved the extraction performance. After functionalized by bio-polydopamine method, strong hydrophobic p-naphtholbenzein molecular was immobilized onto the chemical resistant PEEK surface (PNB@E-PEEK) as stationary phase. We also firstly developed a simple detachable dumbbell-shaped structure for improving the workability of PEEK jacket stir bar. The dumbbell-shaped construction can eliminate the friction between stir bar and container, and the design of detachable structure make elution can be accomplished easier with small amount of organic solvent. It was interesting that the developed detachable dumbbell-shaped PNB@E-PEEK stir bar showed exceptional stability and extraction efficiency for SBSE enrichment of multiple analytes including several Sudan dyes, triazines, polycyclic aromatic hydrocarbons (PAHs), alkaloids and flavonoid. By coupling with high performance liquid chromatography-ultraviolet detection (HPLC-UV), PNB@E-PEEK stir bar based SBSE-HPLC-UV method was applied for the analysis of common Sudan dye pollutants. The method showed low limits of detection (0.02-0.03 ng/mL), good linearity (R2 ≥ 0.9979) and good reproducibility (relative standard deviation ≤ 7.96%). It has been successfully applied to determine three dye pollutants in tap and lake water.