MilliporeSigma
  • Synthesis and characterization of teranthene: a singlet biradical polycyclic aromatic hydrocarbon having Kekule structures.
Synthesis and characterization of teranthene: a singlet biradical polycyclic aromatic hydrocarbon having Kekule structures.
Journal of the American Chemical Society (2010)
Konishi A, et al.