MilliporeSigma
  • Synthesis and Structure of the Thallium (I) Salt of the Tetrakis {3, 5-bis (trifluoromethyl) phenyl} borate Anion.
Synthesis and Structure of the Thallium (I) Salt of the Tetrakis {3, 5-bis (trifluoromethyl) phenyl} borate Anion.
Inorganic Chemistry (1997)
Hughes R P, et al.