MilliporeSigma
  • Chiral 3D architectures with helical channels constructed from polyoxometalate clusters and copper?amino acid complexes.
Chiral 3D architectures with helical channels constructed from polyoxometalate clusters and copper?amino acid complexes.
Angewandte Chemie (International Edition in English) (2006)
An H Y, et al.