MilliporeSigma
  • A Facile and Rapid Route to Highly Enantiopure 1, 2?Diols by Novel Catalytic Asymmetric α?Aminoxylation of Aldehydes.
A Facile and Rapid Route to Highly Enantiopure 1, 2?Diols by Novel Catalytic Asymmetric α?Aminoxylation of Aldehydes.
Angewandte Chemie (International Edition in English) (2003)
Zhong G