MilliporeSigma
  • ATF-2 is a common nuclear target of Smad and TAK1 pathways in transforming growth factor-beta signaling.
ATF-2 is a common nuclear target of Smad and TAK1 pathways in transforming growth factor-beta signaling.
The Journal of biological chemistry (1999-03-20)
Y Sano, J Harada, S Tashiro, R Gotoh-Mandeville, T Maekawa, S Ishii
ABSTRACT
Upon transforming growth factor-beta (TGF-beta) binding to its cognate receptor, Smad3 and Smad4 form heterodimers and transduce the TGF-beta signal to the nucleus. In addition to the Smad pathway, another pathway involving a member of the mitogen-activated protein kinase kinase kinase family of kinases, TGF-beta-activated kinase-1 (TAK1), is required for TGF-beta signaling. However, it is unknown how these pathways function together to synergistically amplify TGF-beta signaling. Here we report that the transcription factor ATF-2 (also called CRE-BP1) is bound by a hetero-oligomer of Smad3 and Smad4 upon TGF-beta stimulation. ATF-2 is one member of the ATF/CREB family that binds to the cAMP response element, and its activity is enhanced after phosphorylation by stress-activated protein kinases such as c-Jun N-terminal kinase and p38. The binding between ATF-2 and Smad3/4 is mediated via the MH1 region of the Smad proteins and the basic leucine zipper region of ATF-2. TGF-beta signaling also induces the phosphorylation of ATF-2 via TAK1 and p38. Both of these actions are shown to be responsible for the synergistic stimulation of ATF-2 trans-activating capacity. These results indicate that ATF-2 plays a central role in TGF-beta signaling by acting as a common nuclear target of both Smad and TAK1 pathways.