MilliporeSigma
  • Comparison of partially and fully chemically-modified siRNA in conjugate-mediated delivery in vivo.
Comparison of partially and fully chemically-modified siRNA in conjugate-mediated delivery <I>in vivo.</I>
Nucleic Acids Research (2018)
Hassler M R, et al.