MilliporeSigma
  • Recapitulating spatiotemporal tumor heterogeneity in vitro through engineered breast cancer microtissues.
Recapitulating spatiotemporal tumor heterogeneity in vitro through engineered breast cancer microtissues.
Acta Biomaterialia (2018)
Mazio C, et al.