MilliporeSigma
  • Natural variant of collagen-like protein A in serotype M3 group A Streptococcus increases adherence and decreases invasive potential.
Natural variant of collagen-like protein A in serotype M3 group A Streptococcus increases adherence and decreases invasive potential.
Infection and Immunity (2015)
Flores A R, et al.