MilliporeSigma
  • Construction and characterization of a multilayered gingival keratinocyte culture model: the TURK-U model.
Construction and characterization of a multilayered gingival keratinocyte culture model: the TURK-U model.
Cytotechnology (2016)
Gursoy U K, et al.