MilliporeSigma
  • Generation of an induced pluripotent stem cell line from chorionic villi of a Turner syndrome spontaneous abortion.
Generation of an induced pluripotent stem cell line from chorionic villi of a Turner syndrome spontaneous abortion.
Stem Cell Research (2017)
Parveen S, et al.