MilliporeSigma
  • Nanoparticle amplification via photothermal unveiling of cryptic collagen binding sites.
Nanoparticle amplification via photothermal unveiling of cryptic collagen binding sites.
Journal of Material Chemistry B: Materials for Biology and Medicine (2013)
Lo J H, et al.