MilliporeSigma
  • Dissolution of fungal cell walls by a streptomycete chitinase and beta-(1-3) glucanase.
Dissolution of fungal cell walls by a streptomycete chitinase and beta-(1-3) glucanase.
Archives of biochemistry and biophysics (1965-08-01)
J J Skujins, H J Potgieter, M Alexander
Sigma-Aldrich
Chitin from shrimp shells, practical grade, powder