MilliporeSigma
  • GSDMD is critical for autoinflammatory pathology in a mouse model of Familial Mediterranean Fever.
GSDMD is critical for autoinflammatory pathology in a mouse model of Familial Mediterranean Fever.
The Journal of experimental medicine (2018-05-26)
Apurva Kanneganti, R K Subbarao Malireddi, Pedro H V Saavedra, Lieselotte Vande Walle, Hanne Van Gorp, Hiroto Kambara, Heather Tillman, Peter Vogel, Hongbo R Luo, Ramnik J Xavier, Hongbo Chi, Mohamed Lamkanfi
ABSTRACT
Pyroptosis is an inflammasome-induced lytic cell death mode, the physiological role of which in chronic inflammatory diseases is unknown. Familial Mediterranean Fever (FMF) is the most common monogenic autoinflammatory disease worldwide, affecting an estimated 150,000 patients. The disease is caused by missense mutations in Mefv that activate the Pyrin inflammasome, but the pathophysiologic mechanisms driving autoinflammation in FMF are incompletely understood. Here, we show that Clostridium difficile infection of FMF knock-in macrophages that express a chimeric FMF-associated MefvV726A Pyrin elicited pyroptosis and gasdermin D (GSDMD)-mediated interleukin (IL)-1β secretion. Importantly, in vivo GSDMD deletion abolished spontaneous autoinflammatory disease. GSDMD-deficient FMF knock-in mice were fully protected from the runted growth, anemia, systemic inflammatory cytokine production, neutrophilia, and tissue damage that characterize this autoinflammatory disease model. Overall, this work identifies pyroptosis as a critical mechanism of IL-1β-dependent autoinflammation in FMF and highlights GSDMD inhibition as a potential antiinflammatory strategy in inflammasome-driven diseases.
Product Description
Sodium taurocholate, BRP, European Pharmacopoeia (EP) Reference Standard