MilliporeSigma
  • Polysorbates 20 and 80 used in the formulation of protein biotherapeutics: structure and degradation pathways.
Polysorbates 20 and 80 used in the formulation of protein biotherapeutics: structure and degradation pathways.
Journal of Pharmaceutical Sciences (2008)
Kerwin BA