MilliporeSigma
  • Arabidopsis thaliana IRX 10 and two related proteins from psyllium and P hyscomitrella patens are xylan xylosyltransferases.
Arabidopsis thaliana IRX 10 and two related proteins from psyllium and P hyscomitrella patens are xylan xylosyltransferases.
The Plant Journal (2014)
Jensen JK, et al.