MilliporeSigma
  • Vimentin is a potential prognostic factor for tongue squamous cell carcinoma among five epithelial?mesenchymal transition-related proteins.
Vimentin is a potential prognostic factor for tongue squamous cell carcinoma among five epithelial?mesenchymal transition-related proteins.
PLoS ONE (2017)
Liu P F, et al.