MilliporeSigma
  • Beta-tubulin binds Src homology 2 domains through a region different from the tyrosine-phosphorylated protein-recognizing site.
Beta-tubulin binds Src homology 2 domains through a region different from the tyrosine-phosphorylated protein-recognizing site.
The Journal of Biological Chemistry (1996)
Itoh T