MilliporeSigma
  • Stereoselective total synthesis of zaragozic acid A based on an acetal [1, 2] Wittig rearrangement.
Stereoselective total synthesis of zaragozic acid A based on an acetal [1, 2] Wittig rearrangement.
Angewandte Chemie (International Edition in English) (2000)
Tomooka, Katsuhiko et al.