MilliporeSigma
  • Design and parallel synthesis of new bicyclic small molecules for targeting the melanocortin receptors.
Design and parallel synthesis of new bicyclic small molecules for targeting the melanocortin receptors.
Advances in experimental medicine and biology (2009-04-30)
Byoung J Min, Alexander V Mayorov, Minying Cai, Erin Palmer, Victor J Hruby
Sigma-Aldrich
Indole-3-propionic acid, ≥97.0% (T)