MilliporeSigma
  • SOK1 translocates from the Golgi to the nucleus upon chemical anoxia and induces apoptotic cell death.
SOK1 translocates from the Golgi to the nucleus upon chemical anoxia and induces apoptotic cell death.
The Journal of biological chemistry (2008-03-28)
Emilio Nogueira, Miguel Fidalgo, Arpad Molnar, John Kyriakis, Thomas Force, Juan Zalvide, Celia M Pombo
ABSTRACT
SOK1 is a Ste20 protein kinase of the germinal center kinase (GCK) family that has been shown to be activated by oxidant stress and chemical anoxia, a cell culture model of ischemia. More recently, it has been shown to be localized to the Golgi apparatus, where it functions in a signaling pathway required for cell migration and polarization. Herein, we demonstrate that SOK1 regulates cell death after chemical anoxia, as its down-regulation by RNA interference enhances cell survival. Furthermore, expression of SOK1 elicits apoptotic cell death by activating the intrinsic pathway. We also find that a cleaved form of SOK1 translocates from the Golgi to the nucleus after chemical anoxia and that this translocation is dependent on both caspase activity and on amino acids 275-292, located immediately C-terminal to the SOK1 kinase domain. Furthermore, SOK1 entry into the nucleus is important for the cell death response since SOK1 mutants unable to enter the nucleus do not induce cell death. In summary, SOK1 is necessary to induce cell death and can induce death when overexpressed. Furthermore, SOK1 appears to play distinctly different roles in stressed versus non-stressed cells, regulating cell death in the former.
Sigma-Aldrich
SOK1, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Golgi FTCD antibody, clone 58k-9, Mouse monoclonal, ascites fluid