- Intestinal secretory defects and dwarfism in mice lacking cGMP-dependent protein kinase II.
Intestinal secretory defects and dwarfism in mice lacking cGMP-dependent protein kinase II.
Science (New York, N.Y.) (1996-12-20)
A Pfeifer, A Aszódi, U Seidler, P Ruth, F Hofmann, R Fässler
PMID8953039
ABSTRACT
Cyclic guanosine 3',5'-monophosphate (cGMP)-dependent protein kinases (cGKs) mediate cellular signaling induced by nitric oxide and cGMP. Mice deficient in the type II cGK were resistant to Escherichia coli STa, an enterotoxin that stimulates cGMP accumulation and intestinal fluid secretion. The cGKII-deficient mice also developed dwarfism that was caused by a severe defect in endochondral ossification at the growth plates. These results indicate that cGKII plays a central role in diverse physiological processes.
Sigma-Aldrich
PRKG2, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Pricing and availability is not currently available.