MilliporeSigma
  • Molecular biology of Philadelphia chromosome in chronic granulocytic leukaemia and acute lymphoblastic leukaemia.
Molecular biology of Philadelphia chromosome in chronic granulocytic leukaemia and acute lymphoblastic leukaemia.
Cytotechnology (1987-10-01)
S Eridani, L M Wiedemann, L C Chan, R G Dalton, K K Karhi
ABSTRACT
In classical t(9;22) translocation, as observed in chronic granulocytic leukemia (CGL), a hybrid DNA unit is produced, including a rearranged PHL gene, previously known as bcr (breakpoint cluster region) plus the translocated c-abl gene from chromosome 9: a hybrid bcr-abl protein, p210 is formed, with increased tyrosine kinase activity. Such DNA rearrangement, with a p210 protein synthesis, is also found in cases of Philadelphia-positive acute lymphoblastic leukemia (ALL), but in apparently similar cases the bcr gene is not rearranged, and a novel p190 abl-related protein can be found; c-abl rearrangement has also been observed.It is thus established that correlations between cytogenetic and molecular events can be found in CGL and ALL, as in other haemopoietic malignancies: translocation and possible rearrangement of the c-abl oncogene seem of particular importance in this case.
Sigma-Aldrich
BCR (1-695), GST tagged human, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Pricing and availability is not currently available.