MilliporeSigma
  • Mutations in the protein kinase A R1alpha regulatory subunit cause familial cardiac myxomas and Carney complex.
Mutations in the protein kinase A R1alpha regulatory subunit cause familial cardiac myxomas and Carney complex.
The Journal of clinical investigation (2000-09-07)
M Casey, C J Vaughan, J He, C J Hatcher, J M Winter, S Weremowicz, K Montgomery, R Kucherlapati, C C Morton, C T Basson
ABSTRACT
Cardiac myxomas are benign mesenchymal tumors that can present as components of the human autosomal dominant disorder Carney complex. Syndromic cardiac myxomas are associated with spotty pigmentation of the skin and endocrinopathy. Our linkage analysis mapped a Carney complex gene defect to chromosome 17q24. We now demonstrate that the PRKAR1alpha gene encoding the R1alpha regulatory subunit of cAMP-dependent protein kinase A (PKA) maps to this chromosome 17q24 locus. Furthermore, we show that PRKAR1alpha frameshift mutations in three unrelated families result in haploinsufficiency of R1alpha and cause Carney complex. We did not detect any truncated R1alpha protein encoded by mutant PRKAR1alpha. Although cardiac tumorigenesis may require a second somatic mutation, DNA and protein analyses of an atrial myxoma resected from a Carney complex patient with a PRKAR1alpha deletion revealed that the myxoma cells retain both the wild-type and the mutant PRKAR1alpha alleles and that wild-type R1alpha protein is stably expressed. However, in this atrial myxoma, we did observe a reversal of the ratio of R1alpha to R2beta regulatory subunit protein, which may contribute to tumorigenesis. Further investigation will elucidate the cell-specific effects of PRKAR1alpha haploinsufficiency on PKA activity and the role of PKA in cardiac growth and differentiation.