MilliporeSigma
  • cDNA cloning and chromosomal mapping of genes encoding novel protein kinases termed PKU-alpha and PKU-beta, which have nuclear localization signal.
cDNA cloning and chromosomal mapping of genes encoding novel protein kinases termed PKU-alpha and PKU-beta, which have nuclear localization signal.
Gene (1998-01-14)
A Yamakawa, Y Kameoka, K Hashimoto, Y Yoshitake, K Nishikawa, K Tanihara, T Date
ABSTRACT
We have cloned cDNAs for novel serine/threonine protein kinases (PK), termed PKU-alpha and PKU-beta, by screening a bacteriophage expression library for kinase activity. Sequence analysis of PKU-alpha and PKU-beta genes revealed that their open reading frames (ORF) were 2151 and 2361 nucleotides (nt) encoding polypeptides of 717 and 787 amino acid (aa) residues, respectively. The deduced aa sequences of PKU-alpha and PKU-beta contained typical serine/threonine PK domains at the C-terminal region and were 86% identical to each other, indicating that they belong to the same PK family. Northern analysis reveals that they are expressed in nearly all human tissues and in cultured cells. The genes for PKU-alpha and PKU-beta were mapped to chromosome 17q23 and 8p12-p22, respectively, by fluorescence in situ hybridization. The proteins encoded by both cDNAs contain a putative nuclear localization signal (NLS) in their N-terminal region. These signals are likely to function in nuclear localization. Glutathione S-transferase (GST)-fusions to regions of PKU-alpha and beta containing the NLS were efficiently localized to the nucleus. In addition, PKU-beta transiently expressed in COS-1 cells was predominantly nuclear. PKU-alpha and PKU-beta differ: a consensus sequence for a nt binding motif is present near the NLS of PKU-beta. These results suggest that PKU-alpha and beta may phosphorylate serine and/or threonine residues on similar proteins, but their activities are regulated through distinct interactions with a nuclear component.