MilliporeSigma
  • Effects of ATP antagonists on purinoceptor-operated inward currents in rat phaeochromocytoma cells.
Effects of ATP antagonists on purinoceptor-operated inward currents in rat phaeochromocytoma cells.
Pflugers Archiv : European journal of physiology (1991-04-01)
K Nakazawa, K Inoue, K Fujimori, A Takanaka
ABSTRACT
The effects of suramin, reactive blue 2 (RB2) and d-tubocurarine (d-TC) were investigated electrophysiologically to elucidate the mechanisms underlying their antagonism of P2 purinoceptor-mediated responses. All three compounds inhibited an adenosine triphosphate (ATP)-activated inward current in rat phaeochromocytoma PC12 cells in a concentration-dependent manner. The order of potency was RB2 greater than suramin greater than d-TC. The inhibition induced by suramin or RB2 was reversible, whereas that induced by d-TC was not reversed after a 5-min rinse. The inactivation of the ATP-activated current was accelerated by d-TC but not by suramin or RB2. RB2 administered simultaneously with ATP exerted much weaker inhibition compared to that induced by prior administration, suggesting that RB2 is a slowly acting antagonist. This was not observed for suramin or d-TC. Suramin and RB2 caused a parallel shift in the concentration/response curve for the ATP-activated current. With d-TC the maximal response of ATP was decreased but the concentration producing half-maximal response was unchanged. The voltage dependency of the ATP-activated current showed less inward rectification in the presence of d-TC. Suramin or RB2 did not affect the voltage dependency. These results suggest that suramin and RB2 reversibly block binding of ATP to receptors, whereas d-TC blocks ion permeability through the ATP-activated channel.