MilliporeSigma
Lipidomics Suggests a New Role for Ceramide Synthase in Phagocytosis.
ACS chemical biology (2018-07-03)
Divya Pathak, Neelay Mehendale, Shubham Singh, Roop Mallik, Siddhesh S Kamat
ABSTRACT
Phagocytosis is an evolutionarily conserved biological process where pathogens or cellular debris are cleared by engulfing them in a membrane-enclosed cellular compartment called the phagosome. The formation, maturation, and subsequent degradation of a phagosome is an important immune response essential for protection against many pathogens. Yet, the global lipid profile of phagosomes remains unknown, especially as a function of their maturation in immune cells. Here, we show using mass spectrometry based quantitative lipidomics that the ceramide class of lipids, especially very long chain ceramides, are enriched on maturing phagosomes with a concomitant decrease in the biosynthetic precursors of ceramides. We thus posit a new function for the enzyme ceramide synthase during phagocytosis in mammalian macrophages. Biochemical assays, cellular lipid feeding experiments, and pharmacological blockade of ceramide synthase together show that this enzyme indeed controls the flux of ceramides on maturing phagosomes. We also find similar results in the primitive eukaryote Dictyostelium discoideum, suggesting that ceramide enrichment may be evolutionarily conserved and likely an indispensible step in phagosome maturation.
Sigma-Aldrich
Anti-CERS2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution