- First synthesis of free cholesterol-BODIPY conjugates.
First synthesis of free cholesterol-BODIPY conjugates.
The Journal of organic chemistry (2006-02-14)
Zaiguo Li, Evan Mintzer, Robert Bittman
PMID16468832
ABSTRACT
Analogues of cholesterol (compounds 1 and 2) and coprostanol (compound 3) containing the BODIPY fluorophore in the aliphatic tail of the free sterol have been synthesized starting with bisnorcholenic acid, cholenic acid 3beta-acetate, and lithocholic acid, respectively. An ester linkage joining the fluorophore to the sterol nucleus interfered with the ability of the fluorescent sterol to pack with phospholipids in monolayers. However, an analogue in which the linker was devoid of polar atoms exhibited a substantially similar physical behavior to cholesterol in model membranes with respect to localization in raft domains.