MilliporeSigma
  • ARTS mediates apoptosis and regeneration of the intestinal stem cell niche.
ARTS mediates apoptosis and regeneration of the intestinal stem cell niche.
Nature communications (2018-11-06)
Elle Koren, Yahav Yosefzon, Roi Ankawa, Despina Soteriou, Avi Jacob, Alexander Nevelsky, Rahamim Ben-Yosef, Gil Bar-Sela, Yaron Fuchs
ABSTRACT
Stem cells (SCs) play a pivotal role in fueling homeostasis and regeneration. While much focus has been given to self-renewal and differentiation pathways regulating SC fate, little is known regarding the specific mechanisms utilized for their elimination. Here, we report that the pro-apoptotic protein ARTS (a Septin4 isoform) is highly expressed in cells comprising the intestinal SC niche and that its deletion protects Lgr5+ and Paneth cells from undergoing apoptotic cell death. As a result, the Sept4/ARTS-/- crypt displays augmented proliferation and, in culture, generates massive cystic-like organoids due to enhanced Wnt/β-catenin signaling. Importantly, Sept4/ARTS-/- mice exhibit resistance against intestinal damage in a manner dependent upon Lgr5+ SCs. Finally, we show that ARTS interacts with XIAP in intestinal crypt cells and that deletion of XIAP can abrogate Sept4/ARTS-/--dependent phenotypes. Our results indicate that intestinal SCs utilize specific apoptotic proteins for their elimination, representing a unique target for regenerative medicine.