MilliporeSigma
  • Self-association and active enzyme forms of Naja naja naja and Crotalus atrox phospholipase A2 studied by analytical ultracentrifugation.
Self-association and active enzyme forms of Naja naja naja and Crotalus atrox phospholipase A2 studied by analytical ultracentrifugation.
Biochemistry (1986-12-02)
T Bukowski, D C Teller
ABSTRACT
The dimerization of phospholipase A2 (PLPA2) from Naja naja naja (Pakistani cobra) and Crotalus atrox (Western Diamondback rattlesnake) has been studied from pH 2.5 to 11 at 20 degrees C in 1 mM CaCl2, 0.21 M ionic strength. For the C. atrox enzyme, it was found necessary to use a combination of sedimentation equilibrium and fluorescence yield data to analyze the association. Sedimentation equilibrium in the analytical ultracentrifuge sufficed for the study of the N. naja PLPA2. In the region of enzymatic activity at pH 8, the dimerization association constants found were k2 = 2.8 X 10(6) L/mol and k2 = 6.9 X 10(4) L/mol for the C. atrox and N. naja enzymes, respectively. Analytical linked functions are presented which describe the data. Because the associations are linked to Ca2+ as well as the hydrogen ion, no attempt was made to interpret the ionization of residues in terms of the molecular structure. Active-enzyme sedimentation velocity experiments have been used to study the relation between enzymatic activity and association for both the C. atrox and N. naja enzymes. The substrate 1,2-dibutyryl-sn-glycero-3-phosphocholine (diC4PC) did not dissociate the C. atrox PLPA2. The substrate 1,2-dihexanoyl-sn-glycero-3-phosphocholine (diC6PC) at 7.5 mM dissociated the C. atrox PLPA2 when monitored either as the active enzyme or as the Sr2+-inhibited enzyme. At low enzyme concentrations, 40 mM diC4PC had no effect on N. naja PLPA2 dimerization. However, the sedimentation coefficients observed at enzyme concentrations above 0.2 mg/mL in active-enzyme sedimentation velocity experiments were larger than the values predicted from the thermodynamic studies. Sedimentation coefficients observed for the N. naja PLPA2 acting on diC6PC were larger than those of the monomeric protein, which was the form layered on this substrate. The dissociation of the C. atrox PLPA2 effected by diC6PC was analyzed by the thermodynamics of association and the kinetic Michaelis constant. The analysis suffices to account for the observed sedimentation coefficient. The sedimentation behavior of the N. naja PLPA2 acting on diC6PC substrate was analyzed in terms of a protein-lipid complex. With this model, 68 +/- 16 phospholipid molecules per protein monomer were determined. It is proposed that this enzyme has two micelle nucleation sites per monomer. These putative sites promote micelle formation of the substrate on the enzyme below the critical micelle concentration of the lipid alone.