MilliporeSigma
  • Uptake of long-chain fatty acids in HepG2 cells involves caveolae: analysis of a novel pathway.
Uptake of long-chain fatty acids in HepG2 cells involves caveolae: analysis of a novel pathway.
Journal of lipid research (2002-09-18)
Jürgen Pohl, Axel Ring, Wolfgang Stremmel
ABSTRACT
We investigated the role of caveolae in uptake and intracellular trafficking of long chain fatty acids (LCFA) in HepG2 human hepatoma cells. The uptake of [(3)H]oleic acid and [(3)H]stearic acid into HepG2 cells was measured by radioactive assays and internalization of the non-metabolizable fluorescent fatty acid 12-(N-methyl)-N-[(7-nitrobenz-2-oxa-1,3-diazol-4-yl)amino] (12-NBD) stearate into single HepG2 cells was semi-quantitatively assessed by laser scanning microscopy. The initial rate of [(3)H]oleic acid uptake (V(0)) in HepG2 cells exhibited saturable transport kinetics with increasing concentrations of free oleic acid (V(max) 854 +/- 46 pmol mg protein(-1) min(-1), K(m) 100 +/- 14 nmol/l). While inhibition of clathrin coated pits did not influence LCFA uptake in HepG2, inhibition of caveolae formation by filipin III, cyclodextrin, and caveolin-1 antisense oligonucleotides resulted in reduction of [(3)H]oleic acid uptake by 54%, 45%, and 23%, respectively. Furthermore, filipin III inhibited the uptake of [(3)H]stearic acid and its fluorescent derivative 12-NBD stearate by 44% and 50%, respectively. Transfection studies with alpha-caveolin-1/cyanofluorescent protein chimeras showed significant colocalization of caveolae and internalized 12-NBD stearate. In conclusion, these data suggest a significant role for caveolae mediated uptake and intracellular trafficking of LCFA in HepG2 cells.