MilliporeSigma
  • Chemical Fingerprinting and Quantification of Chinese Cinnamomi Cortex by Ultra High Performance Liquid Chromatography Coupled with Chemometrics Methods.
Chemical Fingerprinting and Quantification of Chinese Cinnamomi Cortex by Ultra High Performance Liquid Chromatography Coupled with Chemometrics Methods.
Molecules (Basel, Switzerland) (2018-09-12)
Ninghui Ma, Yue Ding, Yong Zhang, Tong Zhang, Yaxiong Yi, Bing Wang
ABSTRACT
To rapidly clarify and quantify the chemical profiling of Cinnamomi cortex a reliable and feasible strategy of chromatographic fingerprinting with a suite of chemometrics methods was developed and validated by ultra-high performance liquid chromatography coupled with diode array detection. Furthermore, to identify more meaningful chemical markers, the chemometrics methods including hierarchical cluster analysis (HCA), principal component analysis (PCA) and similarity, which all generate quality evaluations and correlation classifications of Cinnamomi cortex, were used to improve the Cinnamomi cortex quality control standards. A total of 12 characteristic peaks were confirmed, seven of which were identified by comparing their retention times, UV and MS spectra with authentic compounds. Moreover, 11 analytes were accurately determined, as a complementary quantification method of chromatographic fingerprinting. For quantitative analyses, selective detection was performed at 254, 280 and 340 nm. The tested samples were separated and determined using UPLC and a series of methodologies including linearity, precision, accuracy, limit of detection and quantification and extraction recoveries were validated. Meanwhile the method bias for all the analytes did not exceed 5%. A total of 42 samples were acquired in China and analyzed. The results demonstrated that chromatographic fingerprinting in combination with chemometrics methods provides a promising and practical method to more effectively and comprehensively control the quality of Cinnamomi cortex from various sources, which would be a useful reference for the development and further study of Cinnamomi cortex and related formulations.