MilliporeSigma
  • Ytterbium(III) triflate/TMSCI: efficient catalyst for imino ene reaction
Ytterbium(III) triflate/TMSCI: efficient catalyst for imino ene reaction
Organic letters (2000-05-18)
Yamanaka, Nishida, Nakagawa
ABSTRACT
[reaction: see text] Ytterbium trifluoromethanesulfonate [Yb(OTf)3] catalyzed the imino ene reaction of N-tosyl aldimine with alpha-methylstyrene to give a homoallylamine in moderate yield. Furthermore, addition of a catalytic amount of chlorotrimethylsilane (TMSCI) dramatically enhanced the imino ene reaction.
Sigma-Aldrich
Gadolinium(III) trifluoromethanesulfonate, 98%