MilliporeSigma
  • Nitrile reduction in the presence of Boc-protected amino groups by catalytic hydrogenation over palladium-activated Raney-nickel.
Nitrile reduction in the presence of Boc-protected amino groups by catalytic hydrogenation over palladium-activated Raney-nickel.
The Journal of organic chemistry (2001-04-03)
B Klenke, I H Gilbert
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 3202, slurry, in H2O, active catalyst