MilliporeSigma
  • The potential of hypericin and hyperforin for antiadhesion therapy to prevent metastasis of parental and oxaliplatin-resistant human adenocarcinoma cells (HT-29).
The potential of hypericin and hyperforin for antiadhesion therapy to prevent metastasis of parental and oxaliplatin-resistant human adenocarcinoma cells (HT-29).
Anti-cancer drugs (2018-08-01)
Martina Šemeláková, Veronika Sačková, Peter Fedoročko
ABSTRACT
Cancer cells disseminate to other parts of the body during metastasis through the process of intravasation. The hypericin and hyperforin effect has been described to understand the signal mechanisms that stimulate or stunt cancer cell sprouting to metastasis on colon adenocarcinoma cells HT-29 and its resistant form HT-29-OxR. We focused on the key points of adhesion proteins (cadherin, integrin, selectin and syndecan) and also proteins participating in or contributing to the process of cancer cell migration and adhesion through genes expression and proteins levels. Treatment effects were identified as a consequence of decreased cell adhesion, changes of expression in the adhesive proteins as well as basal membrane degradation associated with changes in the expression of matrix proteinases and in their activity. Finally, the cells affected by hypericin or hyperforin were evaluated by monitoring the cancer cell adhesion properties and proliferation processes. Supplementary Fig. (Supplemental digital content 1, http://links.lww.com/ACD/A267).
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder