MilliporeSigma
  • Human fetal-derived enterospheres provide insights on intestinal development and a novel model to study necrotizing enterocolitis (NEC)
Human fetal-derived enterospheres provide insights on intestinal development and a novel model to study necrotizing enterocolitis (NEC)
Cellular and molecular gastroenterology and hepatology (2018)
Senger S, et al.